
Felbamate

Synonym(s):
2-Phenyl-1,3-propanediol dicarbamate
Empirical Formula (Hill Notation):
C11H14N2O4
CAS Number:
Molecular Weight:
238.24
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing