
Ethylene glycol-bis(succinic acid N-hydroxysuccinimide ester)

Synonym(s):
Di(N-succinimidyl) ethylene glycol disuccinate, EGS, Ethylene glycol bis(succinimidyl succinate), Ethylene glycol disuccinate di(N-succinimidyl) ester
Empirical Formula (Hill Notation):
C18H20N2O12
CAS Number:
Molecular Weight:
456.36
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing