
Doxazosin impurity D

Synonym(s):
6,7-Dimethoxy-2,4-quinazolinedione
Empirical Formula (Hill Notation):
C10H10N2O4
CAS Number:
Molecular Weight:
222.20
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing