Skip to Content
Merck
Dimoxystrobin PESTANAL®, analytical standard

Dimoxystrobin

Dimoxystrobin PESTANAL®, analytical standard
Synonym(s):
(E)-2-(2,5-Dimethylphenoxymethyl)-α-methoxyimino-N-methylphenylacetamide
Empirical Formula (Hill Notation):
C19H22N2O3
CAS Number:
Molecular Weight:
326.39

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique