
Deoxyribonucleic acid, single stranded from calf thymus

Synonym(s):
DNA single stranded
CAS Number:
EC Number:
Products
Product Category
Brand
Physical Form
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing