
Copper bis(2,2,6,6-tetramethyl-3,5-heptanedionate)

Synonym(s):
2,2,6,6-Tetramethyl-3,5-heptanedione copper(II) derivative, Cu(TMHD)2
Linear Formula:
Cu(OCC(CH3)3CHCOC(CH3)3)2
CAS Number:
Molecular Weight:
430.08
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing