
Clozapine for peak identification

Synonym(s):
Clozapine, 8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e][1,4]-diazepine
Empirical Formula (Hill Notation):
C18H19ClN4
CAS Number:
Molecular Weight:
326.82
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing