Skip to Content
IS
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
[β-Mercapto-β,β-cyclopentamethylenepropionyl
1
, O-me-Tyr
2
, Arg
8
]-Vasopressin
Empirical Formula (Hill Notation):
C
52
H
74
N
14
O
12
S
2
CAS Number:
73168-24-8
Molecular Weight:
1151.36
Products
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
V2255
[β-Mercapto-β,β-cyclopentamethylenepropionyl
1
, O-me-Tyr
2
, Arg
8
]-Vasopressin,
≥97% (HPLC)
Expand
View Pricing