
Bicalutamide Related Compound B

Synonym(s):
(RS)-N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(3-fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanamide, Bicalutamide 3-fluoro isomer
Empirical Formula (Hill Notation):
C18H14F4N2O4S
CAS Number:
Molecular Weight:
430.37
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2