
β-Chloro-L-alanine hydrochloride

Empirical Formula (Hill Notation):
C3H6ClNO2 · HCl
CAS Number:
Molecular Weight:
160.00
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing