
Azelastine impurity D

Synonym(s):
4-(4-Chlorobenzyl)phthalazin-1(2H)-one
Empirical Formula (Hill Notation):
C15H11ClN2O
CAS Number:
Molecular Weight:
270.71
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing