
Atractylenolide III

Synonym(s):
8β-Hydroxyasterolide, Codonolactone
Empirical Formula (Hill Notation):
C15H20O3
CAS Number:
Molecular Weight:
248.32
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing