
Ametoctradin

Synonym(s):
5-Ethyl-6-octyl[1,2,4]triazolo[1,5-a]pyrimidin-7-amine
Empirical Formula (Hill Notation):
C15H25N5
CAS Number:
Molecular Weight:
275.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2