
4′-Isobutylacetophenone

Synonym(s):
1-(4-Isobutylphenyl)ethanone, 1-Acetyl-4-isobutylbenzene
Empirical Formula (Hill Notation):
C12H16O
CAS Number:
Molecular Weight:
176.25
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing