
4-Amino-5-Aminomethyl-2-Methylpyrimidine Dihydrochloride

Synonym(s):
4-Amino-5-aminomethyl-2-methylpyrimidine dihydrochloride
Empirical Formula (Hill Notation):
C6H10N4 · 2HCl
CAS Number:
Molecular Weight:
211.09
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing