
3,3′-Diaminobenzidine tetrahydrochloride hydrate

Linear Formula:
C12H14N4 · 4HCl · xH2O
CAS Number:
EC Number:
Products
Product Category
Brand
Color
Physical Form
Purity
Quality Segment
Application
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing