
21-Desacetyl Deflazacort

Synonym(s):
(11β,16β)-11,21-Dihydroxy-2′-methyl-5′H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 21-Desacetyl deflazacort
Empirical Formula (Hill Notation):
C23H29NO5
CAS Number:
Molecular Weight:
399.48
Products
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing