Skip to Content
Merck
All Photos(1)

Key Documents

PHR1357

Supelco

Histamine Dihydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-(4-Imidazolyl)ethylamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. H0600000
traceable to USP 1309009

API family

histamine

CofA

current certificate can be downloaded

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

General description

Histamine Dihydrochloride is a derivative of histamine, which is implicated in tumor cell apoptosis and prevents the relapse of acute myeloid leukemia in patients. It prevents the formation of ROS (reactive oxygen species) and inhibits the activation of T cells and natural killer cells.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA1410 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Markus Lacorn et al.
Journal of AOAC International, 102(5), 1472-1491 (2019-04-25)
Background: RIDASCREEN® Histamine (enzymatic) is an enzymatic test kit for quantification of histamine in fresh fish, canned fish, fish meal, cheese, and wine. Methods: Fish products are extracted with boiling water, while cheese is extracted with water, and the extract

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service