Skip to Content
Merck
All Photos(1)

Key Documents

Z500127

Hose Barb Adapters

polypropylene insert, fits tubing, 1/4 in.

Synonym(s):

hose adapter, hose adapters, hose barbs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.35

material

polypropylene insert

feature

fits tubing 1/4 in.

packaging

pkg of 6 ea

manufacturer/tradename

Nalgene 2159-0001

Looking for similar products? Visit Product Comparison Guide

General description

For use with Nalgene quick filling/venting closures.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service