Skip to Content
Merck
All Photos(1)

Key Documents

CDS013307

Sigma-Aldrich

guaiacylglycerol-beta-guaiacyl ether

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20O6
Molecular Weight:
320.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

SMILES string

COC(C=CC=C1)=C1OC(CO)C(O)C2=CC=C(O)C(OC)=C2

InChI

1S/C17H20O6/c1-21-13-5-3-4-6-14(13)23-16(10-18)17(20)11-7-8-12(19)15(9-11)22-2/h3-9,16-20H,10H2,1-2H3

InChI key

PPZSOILKWHVNNS-UHFFFAOYSA-N

Related Categories

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Qu et al.
ChemSusChem, 13(17), 4510-4518 (2020-04-11)
Model erythro, phenolic, and nonphenolic lignin β-O-4 dimer compounds are treated with copper oxide and H2 O2 at the electronic field maximum position of a single-mode 2.45 GHz microwave system equipped with a cavity resonator. The products obtained through microwave heating
Xinjie Tong et al.
The journal of physical chemistry. B, 123(39), 8247-8260 (2019-09-06)
High resolution differential scanning calorimetry (DSC) and molecular dynamics (MD) simulations were used to investigate the effect of three lignin dimers on the gel to fluid phase transition in DPPC lipid bilayers. The goal of this research is to begin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service