Skip to Content
Merck
All Photos(1)

Key Documents

BMSE1201

Benchmark myGel InstaView Complete Electrophoresis System with Blue LED Illuminator

AC/DC input 115 V

Synonym(s):

protein separation, gel imaging, gel casting, transillumination

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105300
NACRES:
NC.01

manufacturer/tradename

Benchmark Scientific E1201

Looking for similar products? Visit Product Comparison Guide

General description

myGel InstaView Gel Electrophoresis System allows users to view sample migration during electrophoresis. Includes everything needed for casting and running gelx inlcuding gel tank with integrated power supply, casting set, blue LED transilluminator, orange filter cover and imaging enclosure.

Application

Electrophoresis

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service