Skip to Content
Merck
All Photos(1)

Key Documents

603007

Sigma-Aldrich

Water-17O

35-39.9 atom % 17O

Synonym(s):

17O Labeled water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H217O
CAS Number:
Molecular Weight:
19.02
MDL number:
UNSPSC Code:
50202301
PubChem Substance ID:

isotopic purity

35-39.9 atom % 17O

mol wt

18.37 by atom % calculation (at 35 atom % 17O)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.044 g/mL at 25 °C

bulk density

1.01

mass shift

M+1

SMILES string

[17OH2]

InChI

1S/H2O/h1H2/i1+1

InChI key

XLYOFNOQVPJJNP-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(Normalized with respect to hydrogen)

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service