Skip to Content
Merck
All Photos(1)

Key Documents

454699

Sigma-Aldrich

(S)-(+)-Glycidyl methyl ether

97%

Synonym(s):

(S)-(+)-2-(Methoxymethyl)oxirane, (S)-(+)-Methyl glycidyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102505
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +15.6°, c = 5 in toluene

refractive index

n20/D 1.405 (lit.)

bp

110-111 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

SMILES string

COC[C@@H]1CO1

InChI

1S/C4H8O2/c1-5-2-4-3-6-4/h4H,2-3H2,1H3/t4-/m1/s1

InChI key

LKMJVFRMDSNFRT-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

62.6 °F - closed cup

Flash Point(C)

17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service