HPLC Analysis of Pseudoephedrine Enantiomers on Astec® CHIROBIOTIC® TAG
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
[A] methanol; [B] acetic acid; [C] triethylamine;(99.8:0.1:0.1, A:B:C)
flow rate
1 mL/min
detector
UV, 254 nm
injection
10 μL
sample
200 μg/mL in methanol
column temp.
25 °C
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC