HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® V
Materials
analytical column
Application
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] 20 mM sodium citrate; [B] THF; (90:10, A:B)
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 254 nm
injection
10 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC