HPLC Analysis of Ketoprofen Enantiomers on Astec® CHIROBIOTIC® R
Materials
analytical column
CONDITIONS
column
CHIROBIOTIC R, 15 cm x 4.6 mm I.D., 5 μm particles (13023AST)
mobile phase
[A] 20 mM ammonium acetate, pH 5.6; [B] methanol; (50:50, A:B)
flow rate
0.6 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
1 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC