Merck
GC Analysis of Aldehydes (PFBOA Derivatives) in Beer on SLB®-5ms after SPME using a 65 μm PDMS/DVB Fiber
GC Analysis of Aldehydes (PFBOA Derivatives) in Beer on SLB®-5ms after SPME using a 65 μm PDMS/DVB Fiber suitable for GC, application for SPME
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.50 μm
SPME fiber
Product No.
Description
Pricing
SPME Fiber Assembly
65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
related product
Product No.
Description
Pricing
SPME Fiber Holder
for use with Manual Sampling, pk of 1
SPME Fiber Holder
for use with CTC CombiPAL, Gerstel MPS2 Autosamplers, pk of 1
standard
Product No.
Description
Pricing
Benzaldehyde
analytical standard
Phenylacetaldehyde
≥95%, FCC, FG
Isobutyraldehyde
analytical standard
2-Methylbutyraldehyde
analytical standard
Isovaleraldehyde
analytical standard
trans-2-Nonenal
analytical standard
cis-11-Hexadecenal
analytical standard
2-furaldehyde
certified reference material, pharmaceutical secondary standard
CONDITIONS
sample preparation
derivatization procedure: Add 100 μL of PFBOA solution (6 g/L) to 10 mL of deionized water in a 20 mL glass vial and seal. (Insert the PDMS/DVB SPME fiber in the headspace of the PFBOA solution for 10 min at 50 °C.)
sample/matrix
10 mL of degassed beer, 3.5 gm sodium chloride in 20 mL vial
SPME fiber
PDMS-DVB, 23 guage, 65 μm, Auto, Pk/3 (57345-U)
extraction
60 min, headspace, 50 °C, agitation (5 min on, 30 min off)
desorption process
0.10 min, 250 °C
column
SLB-5ms, 30 m x 0.25 mm I.D., 0.50 μm (28473-U)
oven
40 °C, 7 °C/min to 250 °C (14 min)
inj. temp.
250 °C
detector
MSD, SIM
MSD interface
275 °C
carrier gas
helium, 1.1 mL/min
injection
splitless
liner
0.75 mm I.D. SPME liner
sample
Beer
Description
Analysis Note
The formation of aldehydes is a major contributor to the deterioration in flavor or "staling" of beer upon storage. Research indicates that the source of these aldehydes is the type of malt used and quality of the wort produced during the brewing process. This application illustrates the use of SPME to extract aldehydes from beer, followed by on-fiber derivatization with 0-(2,3,4,5,6- pentafluorobenzyl)hydroxylamine hydrochloride (PFBOA). The aldehydes are then analyzed by GC/MS-SIM as their corresponding PFBOA derivatives. The aldehydes exist as geometric isomers, resulting in two derivatives for each. In the case of several of the analytes, these two derivatives were resolved chromatographically.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany