Merck
  • The High-Affinity Interaction between ORC and DNA that Is Required for Replication Licensing Is Inhibited by 2-Arylquinolin-4-Amines.
The High-Affinity Interaction between ORC and DNA that Is Required for Replication Licensing Is Inhibited by 2-Arylquinolin-4-Amines.
Cell chemical biology (2017-08-07)
Nicola J Gardner, Peter J Gillespie, Jamie T Carrington, Emma J Shanks, Stuart P McElroy, Emma J Haagensen, Julie A Frearson, Andrew Woodland, J Julian Blow
ABSTRACT
In late mitosis and G
Sigma-Aldrich
RL5a, ≥98% (HPLC)
Sigma-Aldrich
Octyl-(R)-2HG, ≥98% (HPLC)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture