Merck
Clonally stable Vκ allelic choice instructs Igκ repertoire.
Nature communications (2017-05-31)
Rena Levin-Klein, Shira Fraenkel, Michal Lichtenstein, Louise S Matheson, Osnat Bartok, Yuval Nevo, Sebastian Kadener, Anne E Corcoran, Howard Cedar, Yehudit Bergman
ABSTRACT
Although much has been done to understand how rearrangement of the Igκ locus is regulated during B-cell development, little is known about the way the variable (V) segments themselves are selected. Here we show, using B6/Cast hybrid pre-B-cell clones, that a limited number of V segments on each allele is stochastically activated as characterized by the appearance of non-coding RNA and histone modifications. The activation states are clonally distinct, stable across cell division and developmentally important in directing the Ig repertoire upon differentiation. Using a new approach of allelic ATAC-seq, we demonstrate that the Igκ V alleles have differential chromatin accessibility, which may serve as the underlying basis of clonal maintenance at this locus, as well as other instances of monoallelic expression throughout the genome. These findings highlight a new level of immune system regulation that optimizes gene diversity.
Sigma-Aldrich
Anti-HA Epitope Tag (Influenza Hemaglutinin) Antibody, Chemicon®, from chicken
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit