Merck
  • Ternary Polymer Solar Cells based on Two Acceptors and One Donor for Achieving 12.2% Efficiency.
Ternary Polymer Solar Cells based on Two Acceptors and One Donor for Achieving 12.2% Efficiency.
Advanced materials (Deerfield Beach, Fla.) (2016-11-05)
Wenchao Zhao, Sunsun Li, Shaoqing Zhang, Xiaoyu Liu, Jianhui Hou
ABSTRACT
Ternary polymer solar cells are fabricated based on one donor PBDB-T and two acceptors (a methyl-modified small-molecular acceptor (IT-M) and a bis-adduct of Bis[70]PCBM). A high power conversion efficiency of 12.2% can be achieved. The photovoltaic performance of the ternary polymer solar cells is not sensitive to the composition of the blend.
Sigma-Aldrich
PBDB-T
Pricing and availability is not currently available.