Merck
  • Progressive Muscle Cell Delivery as a Solution for Volumetric Muscle Defect Repair.
Progressive Muscle Cell Delivery as a Solution for Volumetric Muscle Defect Repair.
Scientific reports (2016-12-08)
Ji Hyun Kim, In Kap Ko, Anthony Atala, James J Yoo
ABSTRACT
Reconstructing functional volumetric tissue in vivo following implantation remains a critical challenge facing cell-based approaches. Several pre-vascularization approaches have been developed to increase cell viability following implantation. Structural and functional restoration was achieved in a preclinical rodent tissue defect; however, the approach used in this model fails to repair larger (>mm) defects as observed in a clinical setting. We propose an effective cell delivery system utilizing appropriate vascularization at the site of cell implantation that results in volumetric and functional tissue reconstruction. Our method of multiple cell injections in a progressive manner yielded improved cell survival and formed volumetric muscle tissues in an ectopic muscle site. In addition, this strategy supported the reconstruction of functional skeletal muscle tissue in a rodent volumetric muscle loss injury model. Results from our study suggest that our method may be used to repair volumetric tissue defects by overcoming diffusion limitations and facilitating adequate vascularization.
Sigma-Aldrich
Anti-Nuclei Antibody, clone 235-1, clone 235-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution