Merck
  • Comparison between a chimeric lysin ClyH and other enzymes for extracting DNA to detect methicillin resistant Staphylococcus aureus by quantitative PCR.
Comparison between a chimeric lysin ClyH and other enzymes for extracting DNA to detect methicillin resistant Staphylococcus aureus by quantitative PCR.
World journal of microbiology & biotechnology (2015-11-26)
Yuanyuan Hu, Hang Yang, Jing Wang, Yun Zhang, Junping Yu, Hongping Wei
ABSTRACT
Extracting DNA from Staphylococcus aureus cells is important for detecting MRSA by PCR. However, S. aureus cells are known to be difficult to disrupt due to their compact cell walls. Here, we systematically studied the efficiency of a highly active lysin ClyH for extracting DNA of S. aureus in comparison with commonly used enzymes, such as lysostaphin and achromopeptidase (ACP), and its compatibility in quantitative PCR (qPCR) detection of MRSA. qPCR analysis of S. aureus specific gene femB showed that ClyH was much faster than lysostaphin, ACP and lysozyme for releasing DNA. Five minutes disruption with ClyH at room temperature was enough to release all the DNA from S. aureus. Analysis of the spiked nasal swabs by a dual qPCR assay of the β-lactam resistance mecA gene and the staphylococcal cassette chromosome (SCCmec)-open reading frame X (orfX) junction (SCCmec-orfX) after ClyH lysis showed 100% sensitivity and specificity to the commercial BD GeneOhm™ MRSA test with ACP lysis, but the lysis time was reduced from 20 min by ACP to 5 min by ClyH. Our research shows that ClyH could be a better option than the currently used enzymes for DNA extraction from S. aureus, which can provide simpler and faster PCR detection of MRSA.
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Achromopeptidase from bacteria, lyophilized powder, ≥1,000 units/mg solid
Pricing and availability is not currently available.