Merck
  • An explanation of the noncorrespondence between assessment methods of cyclosporin.
An explanation of the noncorrespondence between assessment methods of cyclosporin.
Transplant international : official journal of the European Society for Organ Transplantation (1989-12-01)
G M Randleff-Rasmussen, M M Oosthuizen, J A Myburgh
ABSTRACT
Various methods of determining cyclosporin (CyA) levels in patients after kidney transplantation were compared. These included polyclonal antibody (pcAb-), specific and nonspecific monoclonal antibody (S- and NmcAb-) radioimmunoassays (RIA), and high performance liquid chromatography (HPLC). The results obtained by the various methods when compared showed some correlation but did not correspond. A probable explanation for part of this noncorrespondence is the presence of monoclonally crossreactive metabolites (CyA-M). Another reason was that the concentration of CyA in the standards supplied with the RIA kits was found to be higher than stated.
Product Description
Supelco
Cyclosporin D solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®