Merck
  • Experimental and computational thermochemical study of N-benzylalanines.
Experimental and computational thermochemical study of N-benzylalanines.
The journal of physical chemistry. B (2011-06-18)
Rafael Notario, Maria Victoria Roux, Concepción Foces-Foces, Manuel A V Ribeiro da Silva, Maria das Dores M C Ribeiro da Silva, Ana Filipa L O M Santos, Ramón Guzmán-Mejía, Eusebio Juaristi
ABSTRACT
Calorimetric measurements are expected to provide useful data regarding the relative stability of α- versus β-amino acid isomers, which, in turn, may help us to understand why nature chose α- instead of β-amino acids for the formation of the biomolecules that are essential constituents of life on earth. The present study is a combination of the experimental determination of the enthalpy of formation of N-benzyl-β-alanine, and high-level ab initio calculations of its molecular structure. The experimentally determined standard molar enthalpy of formation of N-benzyl-β-alanine in gaseous phase at T = 298.15 K is -(298.8 ± 4.8) kJ·mol(-1), whereas its G3(MP2)//B3LYP-calculated enthalpy of formation is -303.7 kJ·mol(-1). This value is in very good agreement with the experimental one. Although the combustion experiments of N-benzyl-α-alanine were unsuccessful, its calculated enthalpy of formation is -310.7 kJ·mol(-1); thus, comparison with the corresponding experimental enthalpy of formation of N-benzyl-β-alanine, -(298.8 ± 4.8) kJ/mol, is in line with the concept that the more branched amino acid (α-alanine) is intrinsically more stable than the linear β-amino acid, β-alanine.
Sigma-Aldrich
L-Homophenylalanine hydrochloride, 97%
Sigma-Aldrich
DL-Homophenylalanine, 98%