Merck
  • Use of a surfactant (polysorbate 80) to improve MIC susceptibility testing results for polymyxin B and colistin.
Use of a surfactant (polysorbate 80) to improve MIC susceptibility testing results for polymyxin B and colistin.
Diagnostic microbiology and infectious disease (2012-10-30)
Helio S Sader, Paul R Rhomberg, Robert K Flamm, Ronald N Jones
ABSTRACT
Accurate determination of in vitro activity for polymyxin class agents has consistently been a problem due to their physical-chemical characteristics that can be influenced by the constituents of reference and/or standardized susceptibility testing methods. We evaluated the impact of using polysorbate 80 (P-80), a surfactant, in reference broth microdilution (BMD) methods when testing polymyxin B and colistin against 247 clinical strains of Enterobacteriaceae (124 strains), Acinetobacter spp. (60 strains), and Pseudomonas aeruginosa (63 strains). All testing was performed in frozen-form BMD panels with and without 0.002% P-80. MIC results for both polymyxins were generally 4- to 8-fold lower when P-80 was added to the testing broth compared to Mueller-Hinton broth without the surfactant. Decreases were greatest in organisms having MIC values at ≤2 μg/mL and among Acinetobacter spp. Polymyxins should be tested with P-80 to more accurately assess the potencies of these agents necessary to treat multidrug-resistant Gram-negative bacilli.
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O
Supelco
Polymyxin B solution, 1 mg/mL in H2O, analytical standard
Sigma-Aldrich
Polymyxin B sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Polymyxin B sulfate salt
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications
Polymyxin B sulfate, European Pharmacopoeia (EP) Reference Standard