Merck
  • Role of detergents in conformational exchange of a G protein-coupled receptor.
Role of detergents in conformational exchange of a G protein-coupled receptor.
The Journal of biological chemistry (2012-08-16)
Ka Young Chung, Tae Hun Kim, Aashish Manglik, Rohan Alvares, Brian K Kobilka, R Scott Prosser
ABSTRACT
The G protein-coupled β(2)-adrenoreceptor (β(2)AR) signals through the heterotrimeric G proteins G(s) and G(i) and β-arrestin. As such, the energy landscape of β(2)AR-excited state conformers is expected to be complex. Upon tagging Cys-265 of β(2)AR with a trifluoromethyl probe, (19)F NMR was used to assess conformations and possible equilibria between states. Here, we report key differences in β(2)AR conformational dynamics associated with the detergents used to stabilize the receptor. In dodecyl maltoside (DDM) micelles, the spectra are well represented by a single Lorentzian line that shifts progressively downfield with activation by appropriate ligand. The results are consistent with interconversion between two or more states on a time scale faster than the greatest difference in ligand-dependent chemical shift (i.e. >100 Hz). Given that high detergent off-rates of DDM monomers may facilitate conformational exchange between functional states of β(2)AR, we utilized the recently developed maltose-neopentyl glycol (MNG-3) diacyl detergent. In MNG-3 micelles, spectra indicated at least three distinct states, the relative populations of which depended on ligand, whereas no ligand-dependent shifts were observed, consistent with the slow exchange limit. Thus, detergent has a profound effect on the equilibrium kinetics between functional states. MNG-3, which has a critical micelle concentration in the nanomolar regime, exhibits an off-rate that is 4 orders of magnitude lower than that of DDM. High detergent off-rates are more likely to facilitate conformational exchange between distinct functional states associated with the G protein-coupled receptor.
Sigma-Aldrich
n-Dodecyl β-D-maltoside, ≥98% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
n-Dodecyl β-D-maltoside, BioXtra, ≥98% (GC)
Pricing and availability is not currently available.