Merck
  • Discovery of γ-secretase modulators with a novel activity profile by text-based virtual screening.
Discovery of γ-secretase modulators with a novel activity profile by text-based virtual screening.
ACS chemical biology (2012-06-26)
Heiko Zettl, Julia Ness, Volker Hähnke, Dirk Beher, Thorsten Jumpertz, Arman Saric, Karlheinz Baumann, Claus U Pietrzik, Bruno Bulic, Gisbert Schneider, Sascha Weggen
ABSTRACT
We present an integrated approach to identify and optimize a novel class of γ-secretase modulators (GSMs) with a unique pharmacological profile. Our strategy included (i) virtual screening through application of a recently developed protocol (PhAST), (ii) synthetic chemistry to discover structure-activity relationships, and (iii) detailed in vitro pharmacological characterization. GSMs are promising agents for treatment or prevention of Alzheimer's disease. They modulate the γ-secretase product spectrum (i.e., amyloid-β (Aβ) peptides of different length) and induce a shift from toxic Aβ42 to shorter Aβ species such as Aβ38 with no or minimal effect on the overall rate of γ-secretase cleavage. We describe the identification of a series of 4-hydroxypyridin-2-one derivatives, which display a novel type of γ-secretase modulation with equipotent inhibition of Aβ42 and Aβ38 peptide species.
Sigma-Aldrich
4-Hydroxypyridine, 95%
Pricing and availability is not currently available.