Merck
  • Toxicological evaluation of ammonium perfluorobutyrate in rats: twenty-eight-day and ninety-day oral gavage studies.
Toxicological evaluation of ammonium perfluorobutyrate in rats: twenty-eight-day and ninety-day oral gavage studies.
Reproductive toxicology (Elmsford, N.Y.) (2011-09-01)
John L Butenhoff, James A Bjork, Shu-Ching Chang, David J Ehresman, George A Parker, Kaberi Das, Christopher Lau, Paul H Lieder, François M van Otterdijk, Kendall B Wallace
ABSTRACT
Sequential 28-day and 90-day oral toxicity studies were performed in male and female rats with ammonium perfluorobutyrate (NH(4)(+)PFBA) at doses up to 150 and 30mg/kg-d, respectively. Ammonium perfluorooctanoate was used as a comparator at a dose of 30mg/kg-d in the 28-day study. Female rats were unaffected by NH(4)(+)PFBA. Effects in males included: increased liver weight, slight to minimal hepatocellular hypertrophy; decreased serum total cholesterol; and reduced serum thyroxin with no change in serum thyrotropin. During recovery, liver weight, histological, and cholesterol effects were resolved. Results of RT-qPCR were consistent with increased transcriptional expression of the xenosensor nuclear receptors PPARα and CAR as well as the thyroid receptor, and decreased expression of Cyp1A1 (Ah receptor-regulated). No observable adverse effect levels (NOAELs) were 6 and >150mg/kg-d for male and female rats in the 28-day study and 6 and >30mg/kg-d in the 90-dat study, respectively.
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Supelco
Heptafluorobutyric acid, suitable for ion chromatography, ≥99.5% (GC)
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)
Supelco
Heptafluorobutyric acid solution, 0.5 M in H2O, LiChropur, suitable for ion pair chromatography