Merck
  • An efficient fluorescence sensor for superoxide with an acridinium ion-linked porphyrin triad.
An efficient fluorescence sensor for superoxide with an acridinium ion-linked porphyrin triad.
Journal of the American Chemical Society (2011-06-28)
Hiroaki Kotani, Kei Ohkubo, Maxwell J Crossley, Shunichi Fukuzumi
ABSTRACT
Addition of potassium superoxide with 18-crown-6 ether (KO(2)(•-)-18-crown-6) to a toluene solution of an acridinium ion-linked porphyrin triad (Acr(+)-H(2)P-Acr(+)) resulted in a remarkable enhancement of the fluorescence intensity. Thus, Acr(+)-H(2)P-Acr(+) acts as an efficient fluorescence sensor for superoxide. Electron transfer from KO(2)(•-)-18-crown-6 to the Acr(+) moiety to produce the two-electron-reduced species (Acr(•)-H(2)P-Acr(•)) results in inhibition of the fluorescence quenching via photoinduced electron transfer, as revealed by laser flash photolysis measurements.
Sigma-Aldrich
18-Crown-6, ≥99.0%
Sigma-Aldrich
18-Crown-6, purum, ≥99.0% (GC)
Sigma-Aldrich
18-Crown-6, 99%
Sigma-Aldrich
18-Crown-6 solution, 1.0 M in THF