Merck
  • Generation of Mouse STO Feeder Cell Lines That Confer Resistance to Several Types of Selective Drugs.
Generation of Mouse STO Feeder Cell Lines That Confer Resistance to Several Types of Selective Drugs.
Cell medicine (2012-05-08)
Issei Saitoh, Masahiro Sato, Yoko Iwase, Emi Inada, Toshiki Nomura, Eri Akasaka, Youichi Yamasaki, Hirofumi Noguchi
ABSTRACT
Feeder cells are generally required for establishment and maintenance of embryonic stem (ES)/induced pluripotent stem (iPS) cells. Increased demands for generation of those cells carrying various types of vectors (i.e., KO vectors and transgenes) also require feeder cells that confer resistance to any types of preexisting selective drugs. Unfortunately, the use of the feeders that are resistant to various drugs appears to be limited to a few laboratories. Here we generated a set of gene-engineered STO feeder cells that confer resistance to several commercially available drugs. The STO cells, which have long been used as a feeder for mouse ES and embryonal carcinoma (EC) cells, were transfected with pcBIH [carrying bleomycin resistance gene (ble) and hygromycin B phosphotransferase gene (Hyg)], pcBIP [carrying ble and puromycin resistance gene (puro)], or pcBSN [carrying ble and neomycin resistance gene (neo)]. The resulting stably transfectants (termed SHB for pcBIH, SPB for pcBIP, and SNB for pcBSN) exhibited bleomycin/hygromycin, bleomycin/puromycin, or bleomycin/neomycin, as expected. The morphology of these cells passaged over 18 generations was indistinguishable from that of parental STO cells. Of isolated clones, the SHB3, SPB3, and SNB2 clones successfully supported the growth of mouse ES cells in an undifferentiated state, when coculture was performed. PCR analysis revealed the presence of the selective markers in these clones, as expected. These SHB3, SPB3, and SNB2 cells will thus be useful for the acquisition and maintenance of genetically manipulated ES/iPS cells.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
COMPLETE ES CELL MEDIUM W/ 15% FBS AND LIF, Ready to use medium formulated with 15% fetal bovine serum (FBS) and mouse leukemia inhibitory factor (LIF). Each lot is qualified for mouse embryonic stem (ES) cell culture.