Merck
  • α-synuclein-lipoprotein interactions and elevated ApoE level in cerebrospinal fluid from Parkinson's disease patients.
α-synuclein-lipoprotein interactions and elevated ApoE level in cerebrospinal fluid from Parkinson's disease patients.
Proceedings of the National Academy of Sciences of the United States of America (2019-07-05)
Wojciech Paslawski, Justyna Zareba-Paslawska, Xiaoqun Zhang, Katharina Hölzl, Henrik Wadensten, Mohammadreza Shariatgorji, Shorena Janelidze, Oskar Hansson, Lars Forsgren, Per E Andrén, Per Svenningsson
ABSTRACT
The progressive accumulation, aggregation, and spread of α-synuclein (αSN) are common hallmarks of Parkinson's disease (PD) pathology. Moreover, numerous proteins interact with αSN species, influencing its toxicity in the brain. In the present study, we extended analyses of αSN-interacting proteins to cerebrospinal fluid (CSF). Using coimmunoprecipitation, followed by mass spectrometry, we found that αSN colocalize with apolipoproteins on lipoprotein vesicles. We confirmed these interactions using several methods, including the enrichment of lipoproteins with a recombinant αSN, and the subsequent uptake of prepared vesicles by human dopaminergic neuronal-like cells. Further, we report an increased level of ApoE in CSF from early PD patients compared with matched controls in 3 independent cohorts. Moreover, in contrast to controls, we observed the presence of ApoE-positive neuromelanin-containing dopaminergic neurons in substantia nigra of PD patients. In conclusion, the cooccurrence of αSN on lipoprotein vesicles, and their uptake by dopaminergic neurons along with an increase of ApoE in early PD, proposes a mechanism(s) for αSN spreading in the extracellular milieu of PD.
Sigma-Aldrich
Lipoproteins, High Density, Human Plasma
Pricing and availability is not currently available.
Sigma-Aldrich
Lipoproteins, Very Low Density, Human Plasma
Pricing and availability is not currently available.