Merck
  • Targeting nuclear β-catenin as therapy for post-myeloproliferative neoplasm secondary AML.
Targeting nuclear β-catenin as therapy for post-myeloproliferative neoplasm secondary AML.
Leukemia (2018-12-24)
Dyana T Saenz, Warren Fiskus, Taghi Manshouri, Christopher P Mill, Yimin Qian, Kanak Raina, Kimal Rajapakshe, Cristian Coarfa, Raffaella Soldi, Prithviraj Bose, Gautam Borthakur, Tapan M Kadia, Joseph D Khoury, Lucia Masarova, Agnieszka J Nowak, Baohua Sun, David N Saenz, Steven M Kornblau, Steve Horrigan, Sunil Sharma, Peng Qiu, Craig M Crews, Srdan Verstovsek, Kapil N Bhalla
ABSTRACT
Transformation of post-myeloproliferative neoplasms into secondary (s) AML exhibit poor clinical outcome. In addition to increased JAK-STAT and PI3K-AKT signaling, post-MPN sAML blast progenitor cells (BPCs) demonstrate increased nuclear β-catenin levels and TCF7L2 (TCF4) transcriptional activity. Knockdown of β-catenin or treatment with BC2059 that disrupts binding of β-catenin to TBL1X (TBL1) depleted nuclear β-catenin levels. This induced apoptosis of not only JAKi-sensitive but also JAKi-persister/resistant post-MPN sAML BPCs, associated with attenuation of TCF4 transcriptional targets MYC, BCL-2, and Survivin. Co-targeting of β-catenin and JAK1/2 inhibitor ruxolitinib (rux) synergistically induced lethality in post-MPN sAML BPCs and improved survival of mice engrafted with human sAML BPCs. Notably, co-treatment with BET protein degrader ARV-771 and BC2059 also synergistically induced apoptosis and improved survival of mice engrafted with JAKi-sensitive or JAKi-persister/resistant post-MPN sAML cells. These preclinical findings highlight potentially promising anti-post-MPN sAML activity of the combination of β-catenin and BETP antagonists against post-MPN sAML BPCs.
Sigma-Aldrich
ARV-771, ≥98% (HPLC)
Sigma-Aldrich
BC-2059, ≥98% (HPLC)