Merck
  • Genetic interaction between central pair apparatus genes CFAP221, CFAP54, and SPEF2 in mouse models of primary ciliary dyskinesia.
Genetic interaction between central pair apparatus genes CFAP221, CFAP54, and SPEF2 in mouse models of primary ciliary dyskinesia.
Scientific reports (2020-07-25)
Casey W McKenzie, Lance Lee
ABSTRACT
Primary ciliary dyskinesia (PCD) is a genetically heterogeneous syndrome that results from defects in motile cilia. The ciliary axoneme has a 9 + 2 microtubule structure consisting of nine peripheral doublets surrounding a central pair apparatus (CPA), which plays a critical role in regulating proper ciliary function. We have previously shown that mouse models with mutations in CPA genes CFAP221, CFAP54, and SPEF2 have a PCD phenotype with defects in ciliary motility. In this study, we investigated potential genetic interaction between these CPA genes by generating each combination of double heterozygous and double homozygous mutants. No detectable cilia-related phenotypes were observed in double heterozygotes, but all three double homozygous mutant lines exhibit early mortality and typically develop severe PCD-associated phenotypes of hydrocephalus, mucociliary clearance defects in the upper airway, and abnormal spermatogenesis. Double homozygous cilia are generally intact and display a normal morphology and distribution. Spermiogenesis is aborted in double homozygotes, with an absence of mature flagella on elongating spermatids and epididymal sperm. These findings identify genetic interactions between CPA genes and genetic mechanisms regulating the CPA and motile cilia function.
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Anti-DNAI1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Sigma-Aldrich
Anti-RSPH4A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution