Merck
  • Targeting CMTM6 Suppresses Stem Cell-Like Properties and Enhances Antitumor Immunity in Head and Neck Squamous Cell Carcinoma.
Targeting CMTM6 Suppresses Stem Cell-Like Properties and Enhances Antitumor Immunity in Head and Neck Squamous Cell Carcinoma.
Cancer immunology research (2019-11-28)
Lei Chen, Qi-Chao Yang, Yi-Cun Li, Lei-Lei Yang, Jian-Feng Liu, Hao Li, Yao Xiao, Lin-Lin Bu, Wen-Feng Zhang, Zhi-Jun Sun
ABSTRACT
CMTM6, a regulator of PD-L1 expression, also modulates tumor immunity. Little is known about the function of CMTM6 and its mechanism of action in head and neck squamous cell carcinoma (HNSCC). In this study, we found by IHC analysis that CMTM6 overexpression predicted a poor prognosis for patients with HNSCC. We discovered that CMTM6 expression was correlated with increased activity through the Wnt/β-catenin signaling pathway, which is essential for tumorigenesis, maintenance of cancer stem cells (CSC), and the epithelial-to-mesenchymal transition (EMT) characteristic of multiple cancers. We used short hairpin RNA to eliminate expression of CMTM6, which led, in HNSCC cells, to reduced expression of nuclear β-catenin as well as inhibition of stem cell-like properties, TGFβ-induced EMT, and cell proliferation. Consistent with these results, we identified a significant positive correlation between expression of CMTM6 and EMT- and CSC-related genes in The Cancer Genome Atlas (TCGA). We found positive correlations for both RNA and protein between expression of CMTM6 and immune checkpoint components. CMTM6 silencing-induced PD-L1 downregulation delayed SCC7 tumor growth and increased CD8+ and CD4+ T-cell infiltration. The proportions of PD-1+, TIM-3+, VISTA+, LAG-3+, and B7-H3+ exhausted T cells were decreased significantly in the CMTM6 knockdown group. CMTM6 thus regulates stemness, EMT, and T-cell dysfunction and may be a promising therapeutic target in the treatment of HNSCC.
Sigma-Aldrich
Anti-CMTM6 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-CMTM6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution