Merck
  • Deletion of NLRX1 increases fatty acid metabolism and prevents diet-induced hepatic steatosis and metabolic syndrome.
Deletion of NLRX1 increases fatty acid metabolism and prevents diet-induced hepatic steatosis and metabolic syndrome.
Biochimica et biophysica acta. Molecular basis of disease (2018-03-08)
Lotte Kors, Elena Rampanelli, Geurt Stokman, Loes M Butter, Ntsiki M Held, Nike Claessen, Per W B Larsen, Joanne Verheij, Coert J Zuurbier, Gerhard Liebisch, Gerd Schmitz, Stephen E Girardin, Sandrine Florquin, Riekelt H Houtkooper, Jaklien C Leemans
ABSTRACT
NOD-like receptor (NLR)X1 (NLRX1) is an ubiquitously expressed inflammasome-independent NLR that is uniquely localized in mitochondria with as yet unknown effects on metabolic diseases. Here, we report that NLRX1 is essential in regulating cellular metabolism in non-immune parenchymal hepatocytes by decreasing mitochondrial fatty acid-dependent oxidative phosphorylation (OXPHOS) and promoting glycolysis. NLRX1 loss in mice has a profound impact on the prevention of diet-induced metabolic syndrome parameters, non-alcoholic fatty liver disease (NAFLD) progression, and renal dysfunction. Despite enhanced caloric intake, NLRX1 deletion in mice fed a western diet (WD) results in protection from liver steatosis, hepatic fibrosis, obesity, insulin resistance, glycosuria and kidney dysfunction parameters independent from inflammation. While mitochondrial content was equal, NLRX1 loss in hepatocytes leads to increased fatty acid oxidation and decreased steatosis. In contrast, glycolysis was decreased in NLRX1-deficient cells versus controls. Thus, although first implicated in immune regulation, we show that NLRX1 function extends to the control of hepatocyte energy metabolism via the restriction of mitochondrial fatty acid-dependent OXPHOS and enhancement of glycolysis. As such NLRX1 may be an attractive novel therapeutic target for NAFLD and metabolic syndrome.