
Tris Acetate-EDTA buffer

Synonym(s):
TAE buffer
Products
Product Category
Brand
Feature
pH Value
Physical Form
Product Line
Research Area
Quality Segment
Application
Sterility
Sterilization Method
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing