
Tazobactam

Synonym(s):
[2S-(2α,3β,5α)]-3-Methyl-7-oxo-3-(1H,2,3-triazol-1-ylmethyl)-4-thia-1-azabicyclo[3.2.0]heptan-2-carbonsäure-4,4-dioxid, Tazobactam
Empirical Formula (Hill Notation):
C10H12N4O5S
CAS Number:
Molecular Weight:
300.29
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing