
Sesame Oil Related Compound A

Synonym(s):
1,2-Dilinoleoyl-3-oleoyl-rac-glycerol, 1,2-Di(cis-9,12-octadecadienoyl)-3-(cis-9-octadecenoyl)-rac-glycerol, OLL
Empirical Formula (Hill Notation):
C57H100O6
CAS Number:
Molecular Weight:
881.40
Products
Formula Weight
Manufacturer
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing