
Saikosaponin A

Empirical Formula (Hill Notation):
C42H68O13
CAS Number:
Molecular Weight:
780.98
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing