
Rubiadin

Synonym(s):
1,3-Dihydroxy-2-methyl-9,10-anthracenedione, 1,3-Dihydroxy-2-methylanthraquinone
Empirical Formula (Hill Notation):
C15H10O4
CAS Number:
Molecular Weight:
254.24
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing